2-Oxoindoline-3-carbaldehyde, Technical Grade

Supplier: Apollo Scientific
Warning

OR926453-1G OR926453-5G
APOSOR926453-1GEA 49.3 EUR
APOSOR926453-1G APOSOR926453-5G
2-Oxoindoline-3-carbaldehyde, Technical Grade
2-Oxoindoline-3-carbaldehyde
Formula: C₉H₇NO₂
Storage Temperature: Ambient
MDL Number: MFCD00463399
CAS Number: 78610-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR