2-Methyl-3-hexanone 95%
Supplier: Apollo Scientific
OR928462-10G OR928462-1G OR928462-5G
APOSOR928462-10GEA 377 EUR
APOSOR928462-10G APOSOR928462-1G APOSOR928462-5G
2-Methyl-3-hexanone 95%
2-Methyl-3-hexanone
Formula: C₇H₁₄O
MW: 114.19 g/mol
Density: 0.8428
Storage Temperature: Ambient
MDL Number: MFCD00009399
CAS Number: 7379-12-6