tert-Butyl methyl(2-oxoethyl)carbamate 95%
Supplier: Apollo Scientific
Synonyms: tert-Butyl methyll(2-oxoethyl)carbamate, Methyl(2-oxoethyl)carbamic acid tert-butyl ester, N-Boc-(methylamino)acetaldehyde, N-tert-Butoxycarbonyl-(methylamino)acetaldehyde
OR928760-10G OR928760-1G OR928760-5G
APOSOR928760-10GEA 142 EUR
APOSOR928760-10G APOSOR928760-1G APOSOR928760-5G
tert-Butyl methyl(2-oxoethyl)carbamate 95%
tert-Butyl methyl(2-oxoethyl)carbamate
Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08064267
CAS Number: 123387-72-4