2-(Chloromethyl)-1,3-dioxolane 98%
Supplier: Apollo Scientific
OR928993-100G OR928993-25G
APOSOR928993-100GEA 0 EUR
APOSOR928993-100G APOSOR928993-25G
2-(Chloromethyl)-1,3-dioxolane 98%
2-(Chloromethyl)-1,3-dioxolane
Formula: C₄H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00043402
CAS Number: 2568-30-1