Boc-N-ME-PHG-OH 97%
Supplier: Apollo Scientific
Synonyms: N-Boc-N-methyl-L-phenylglycine, Boc-N-Me-Phg-OH
OR929168-1G OR929168-5G
APOSOR929168-1GEA 81.2 EUR
APOSOR929168-1G APOSOR929168-5G
Boc-N-ME-PHG-OH 97%
Boc-N-ME-PHG-OH
Formula: C₁₄H₁₉NO₄
MW: 265.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038799
CAS Number: 30925-11-2