4-Chloro-3,5-xylenol 98%
Supplier: Apollo Scientific
Synonyms: 2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol
OR9315-25G OR9315-500G
APOSOR9315-25GEA 21.9 EUR
APOSOR9315-25G APOSOR9315-500G
4-Chloro-3,5-xylenol 98%
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002324
CAS Number: 88-04-0