2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Supplier: Apollo Scientific
Warning
Synonyms: Benzylboronic Acid Pinacol Ester
OR9316-1G OR9316-5G
APOSOR9316-1GEA 21.9 EUR
APOSOR9316-1G APOSOR9316-5G
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-Benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₉BO₂
Boiling Pt: 65 °C/0.15mmHg
Density: 0.98 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD05663841
CAS Number: 87100-28-5