Methyl-1-naphthaleneacetate 97%
Supplier: Apollo Scientific
OR931999-100G OR931999-25G
APOSOR931999-100GEA 62.5 EUR
APOSOR931999-100G APOSOR931999-25G
Methyl-1-naphthaleneacetate 97%
Methyl-1-naphthaleneacetate
Formula: C₁₃H₁₂O₂
Boiling Pt: 160…162 °C/5.25mmHg
Density: 1.142 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00004045
CAS Number: 2876-78-0