3,4-Dichlorophenylhydrazinium chloride 97%
Supplier: Apollo Scientific
Warning
OR9320-25G OR9320-100G
APOSOR9320-25GEA 31.9 EUR
APOSOR9320-25G APOSOR9320-100G
3,4-Dichlorophenylhydrazinium chloride 97%
3,4-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012937
CAS Number: 19763-90-7
EINECS: 243-278-0