4-Ethyl-1-octyn-3-ol 95%
Supplier: Apollo Scientific
OR932328-25G OR932328-100G
APOSOR932328-25GEA 145 EUR
APOSOR932328-25G APOSOR932328-100G
4-Ethyl-1-octyn-3-ol 95%
4-Ethyl-1-octyn-3-ol
Formula: C₁₀H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00015262
CAS Number: 5877-42-9