N-(Trimethylsilyl)morpholine 97%
Supplier: Apollo Scientific
Synonyms: 4-(Trimethylsilyl)morpholine
OR936934-25G OR936934-100G
APOSOR936934-25GEA 102 EUR
APOSOR936934-25G APOSOR936934-100G
N-(Trimethylsilyl)morpholine 97%
N-(Trimethylsilyl)morpholine
Formula: C₇H₁₇NOSi
MW: 159.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010098
CAS Number: 13368-42-8