4'-Aminohexanophenone 95%

Supplier: Apollo Scientific

OR937047-10G OR937047-1G OR937047-5G
APOSOR937047-10GEA 754 EUR
APOSOR937047-10G APOSOR937047-1G APOSOR937047-5G
4'-Aminohexanophenone 95%
4'-Aminohexanophenone
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
CAS Number: 38237-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR