cis-4-Hepten-1-ol 95%
Supplier: Apollo Scientific
OR937449-10G OR937449-1G OR937449-5G
APOSOR937449-10GEA 406 EUR
APOSOR937449-10G APOSOR937449-1G APOSOR937449-5G
cis-4-Hepten-1-ol 95%
cis-4-Hepten-1-ol
Formula: C₇H₁₄O
MW: 114.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015575
CAS Number: 6191-71-5