1,2-Dihydroxyindan 95%
Supplier: Apollo Scientific
OR937916-1G
APOSOR937916-1GEA 145 EUR
APOSOR937916-1G
1,2-Dihydroxyindan 95%
1,2-Dihydroxyindan
Formula: C₉H₁₀O₂
Storage Temperature: Ambient
CAS Number: 4370-02-9