3-(Cyanomethyl)-2,4,5-trimethylthiophene 95%
Supplier: Apollo Scientific
Synonyms: 2-(2,4,5-Trimethylthiophen-3-yl)acetonitrile
OR938038-1G OR938038-250MG
APOSOR938038-1GEA 102 EUR
APOSOR938038-1G APOSOR938038-250MG
3-(Cyanomethyl)-2,4,5-trimethylthiophene 95%
3-(Cyanomethyl)-2,4,5-trimethylthiophene
Formula: C₉H₁₁NS
MW: 165.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00143113
CAS Number: 112440-49-0