4-Methoxy-3-(4-morpholinylsulphonyl)benzeneboronic acid 95%
Supplier: Apollo Scientific
Warning
OR9390-1G OR9390-500MG
APOSOR9390-1GEA 306 EUR
APOSOR9390-1G APOSOR9390-500MG
4-Methoxy-3-(4-morpholinylsulphonyl)benzeneboronic acid 95%
4-Methoxy-3-(4-morpholinylsulphonyl)benzeneboronic acid
Formula: C₁₁H₁₆BNO₆S
MW: 301.12 g/mol
Melting Pt: 270…276 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056366
CAS Number: 871333-02-7