(4-Chloro-3-cyanophenyl)boronic acid 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-3-cyanophenylboronic acid, 5-Borono-2-chlorobenzonitrile
OR9397-1G OR9397-5G OR9397-10G
APOSOR9397-1GEA 21.9 EUR
APOSOR9397-1G APOSOR9397-5G APOSOR9397-10G
(4-Chloro-3-cyanophenyl)boronic acid 97+%
(4-Chloro-3-cyanophenyl)boronic acid
Formula: C₇H₅BClNO₂
MW: 181.382 g/mol
Melting Pt: 300…304 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056360
CAS Number: 871332-95-5