N-Butylthiophosphoric triamide

Supplier: Apollo Scientific
Warning

Synonyms: N-Butylphosphorothioic triamide

OR9400T-100G OR9400T-25G OR9400T-5G
APOSOR9400T-100GEA 56.7 EUR
APOSOR9400T-100G APOSOR9400T-25G APOSOR9400T-5G
N-Butylthiophosphoric triamide
N-Butylthiophosphoric triamide
Formula: C₄H₁₄N₃PS
Melting Pt: 56…60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00269941
CAS Number: 94317-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR