4-Cyclohexylbutyric acid 95%

Supplier: Apollo Scientific

OR940544-25G OR940544-5G
APOSOR940544-25GEA 128 EUR
APOSOR940544-25G APOSOR940544-5G
4-Cyclohexylbutyric acid 95%
4-Cyclohexylbutyric acid
Formula: C₁₀H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00001529
CAS Number: 4441-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR