2-Nitro-1-phenylpropene 98%
Supplier: Apollo Scientific
Warning
Synonyms: trans-β-Methyl-β-nitrostyrene
OR942029-25G OR942029-5G OR942029-100G
APOSOR942029-25GEA 241 EUR
APOSOR942029-25G APOSOR942029-5G APOSOR942029-100G
2-Nitro-1-phenylpropene 98%
2-Nitro-1-phenylpropene
Formula: C₉H₉NO₂
Melting Pt: 64…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00014720
CAS Number: 705-60-2