2-Methylanisole 99%
Supplier: Apollo Scientific
Warning
Synonyms: o-Cresol methyll ether, 2-Methylanisole, 1-Methoxy-2-methylbenzene, 2-Methoxytoluene
OR9430-250G OR9430-50G
APOSOR9430-250GEA 207 EUR
APOSOR9430-250G APOSOR9430-50G
2-Methylanisole 99%
2-Methylanisole
Formula: C₈H₁₀O
Boiling Pt: 170…172 °C
Melting Pt: –34 °C
Density: 0.985 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00008373
CAS Number: 578-58-5
EINECS: 209-426-3
UN: 3271
ADR: 3,III