tert-Butyl allyl(prop-2-yn-1-yl)carbamate 95%
Supplier: Apollo Scientific
OR943285-1G OR943285-250MG
APOSOR943285-1GEA 770 EUR
APOSOR943285-1G APOSOR943285-250MG
tert-Butyl allyl(prop-2-yn-1-yl)carbamate 95%
tert-Butyl allyl(prop-2-yn-1-yl)carbamate
Formula: C₁₁H₁₇NO₂
MW: 195.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18432512
CAS Number: 147528-20-9