Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid 95%
Supplier: Apollo Scientific
OR944579-1G OR944579-250MG
APOSOR944579-1GEA 406 EUR
APOSOR944579-1G APOSOR944579-250MG
Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid 95%
Pyrazolo[1,5-a]pyrimidine-5-carboxylic acid
Formula: C₇H₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD09414725
CAS Number: 1086375-50-9