2-Bromo-3-methylbenzaldehyde 95%

Supplier: Apollo Scientific

OR944617-1G OR944617-250MG
APOSOR944617-1GEA 0 EUR
APOSOR944617-1G APOSOR944617-250MG
2-Bromo-3-methylbenzaldehyde 95%
2-Bromo-3-methylbenzaldehyde
Formula: C₈H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD18392002
CAS Number: 109179-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR