1,2-Cyclopentane diformic anhydride 98%
Supplier: Apollo Scientific
Synonyms: Tetrahydro-1H-cyclopenta[c]furan-1,3(3ah)-dione
OR945919-1G OR945919-5G
APOSOR945919-1GEA 65.4 EUR
APOSOR945919-1G APOSOR945919-5G
1,2-Cyclopentane diformic anhydride 98%
1,2-Cyclopentane diformic anhydride
Formula: C₇H₈O₃
MW: 140.138 g/mol
Storage Temperature: Ambient
CAS Number: 5763-49-5