1-Isopropyl-1H-1,2,4-triazole 95%

Supplier: Apollo Scientific

OR945994-1G OR945994-250MG
APOSOR945994-1GEA 647 EUR
APOSOR945994-1G APOSOR945994-250MG
1-Isopropyl-1H-1,2,4-triazole 95%
1-Isopropyl-1H-1,2,4-triazole
Formula: C₅H₉N₃
Storage Temperature: Ambient
MDL Number: MFCD18889704
CAS Number: 63936-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR