3-Bromo-4-methylthiophene 97%
Supplier: Apollo Scientific
Warning
OR9461-1G OR9461-25G OR9461-5G
APOSOR9461-1GEA 40.8 EUR
APOSOR9461-1G APOSOR9461-25G APOSOR9461-5G
3-Bromo-4-methylthiophene 97%
3-Bromo-4-methylthiophene
Formula: C₅H₅BrS
Boiling Pt: 179…183 °C
Density: 1.584 g/cm³ (25 °C)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130084
CAS Number: 30318-99-1