3,4-Dichlorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3,4-Dichlorobenzene-1-sulphonyl chloride
OR9483-100G OR9483-25G OR9483-5G
APOSOR9483-100GEA 87 EUR
APOSOR9483-100G APOSOR9483-25G APOSOR9483-5G
3,4-Dichlorobenzenesulphonyl chloride 98%
3,4-Dichlorobenzenesulphonyl chloride
Formula: C₆H₃Cl₃O₂S
Boiling Pt: 253 °C
Density: 1.572 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00041255
CAS Number: 98-31-7
EINECS: 202-656-5
UN: 2920
ADR: 8+3,II