3,6-Dimethyl-4-octyne-3,6-diol 95%
Supplier: Apollo Scientific
Synonyms: 3,6-Dihydroxy-3,6-dimethyl-4-octyne
OR950160-25G OR950160-100G OR950160-500G OR950160-5G
APOSOR950160-25GEA 116 EUR
APOSOR950160-25G APOSOR950160-100G APOSOR950160-500G APOSOR950160-5G
3,6-Dimethyl-4-octyne-3,6-diol 95%
3,6-Dimethyl-4-octyne-3,6-diol
Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004480
CAS Number: 78-66-0