2-(4-Pyridyl)-2-propanol 97%
Supplier: Apollo Scientific
Synonyms: 2-(4-Pyridyl)isopropyl Alcohol
OR951271-25G OR951271-5G
APOSOR951271-25GEA 81.2 EUR
APOSOR951271-25G APOSOR951271-5G
2-(4-Pyridyl)-2-propanol 97%
2-(4-Pyridyl)-2-propanol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071746
CAS Number: 15031-78-4