2-Pyrimidineacetic acid 98%
Supplier: Apollo Scientific
Warning
OR951464-1G OR951464-250MG OR951464-5G
APOSOR951464-1GEA 84.1 EUR
APOSOR951464-1G APOSOR951464-250MG APOSOR951464-5G
2-Pyrimidineacetic acid 98%
2-Pyrimidineacetic acid
Formula: C₆H₆N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD04973302
CAS Number: 66621-73-6