4-Bromo-6-chloro-1H-indazole 98%
Supplier: Apollo Scientific
Warning
OR951968-10G OR951968-1G OR951968-5G
APOSOR951968-10GEA 226 EUR
APOSOR951968-10G APOSOR951968-1G APOSOR951968-5G
4-Bromo-6-chloro-1H-indazole 98%
4-Bromo-6-chloro-1H-indazole
Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781352
CAS Number: 885519-03-9