7-Methoxy-1,4-benzothiazin-3-one 95%
Supplier: Apollo Scientific
OR952627-10G OR952627-1G OR952627-5G
APOSOR952627-10GEA 696 EUR
APOSOR952627-10G APOSOR952627-1G APOSOR952627-5G
7-Methoxy-1,4-benzothiazin-3-one 95%
7-Methoxy-1,4-benzothiazin-3-one
Formula: C₉H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD02660686
CAS Number: 22726-30-3