Methyl-3-(benzyloxy)-4-bromobenzoate 98%
Supplier: Apollo Scientific
Warning
OR9535-1G OR9535-250MG
APOSOR9535-1GEA 170 EUR
APOSOR9535-1G APOSOR9535-250MG
Methyl-3-(benzyloxy)-4-bromobenzoate 98%
Methyl 3-(benzyloxy)-4-bromobenzoate
Formula: C₁₅H₁₃BrO₃
Boiling Pt: 202…203 °C/4mmHg
Melting Pt: 81…83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08436016
CAS Number: 17054-26-1