[2-(3-Methylphenoxy)ethyl]amine hydrochloride 95%
Supplier: Apollo Scientific
Synonyms: 2-(3-Methylphenoxy)ethanamine
OR953999-1G OR953999-250MG
APOSOR953999-1GEA 140 EUR
APOSOR953999-1G APOSOR953999-250MG
[2-(3-Methylphenoxy)ethyl]amine hydrochloride 95%
[2-(3-Methylphenoxy)ethyl]amine hydrochloride
Formula: C₉H₁₄ClNO
MW: 187.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02667723
CAS Number: 6487-99-6