2,3-Dichloropyrazine 98%
Supplier: Apollo Scientific
Warning
OR9580-100G OR9580-25G
APOSOR9580-100GEA 42.2 EUR
APOSOR9580-100G APOSOR9580-25G
2,3-Dichloropyrazine 98%
2,3-Dichloropyrazine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 105…107 °C/50mmHg
Melting Pt: 22.5…24.5 °C
Density: 1.431 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00040964
CAS Number: 4858-85-9
EINECS: 225-460-1