1-Bromo-4-iodobenzene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-1-iodobenzene
OR9587-100G OR9587-25G OR9587-500G
APOSOR9587-100GEA 53.8 EUR
APOSOR9587-100G APOSOR9587-25G APOSOR9587-500G
1-Bromo-4-iodobenzene 98%
1-Bromo-4-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 120…122 °C/14mmHg
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00001051
CAS Number: 589-87-7
EINECS: 209-662-7