2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Acetyl-5,5-dimethylcyclohexane-1,3-dione, 2-Acetyldimedone
OR962381-25G OR962381-5G
APOSOR962381-25GEA 330 EUR
APOSOR962381-25G APOSOR962381-5G
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98%
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione
Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
Melting Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00467663
CAS Number: 1755-15-3
EINECS: 626-974-2