Ac-Nva-OH 98%
Supplier: Apollo Scientific
OR963214-25G OR963214-5G OR963214-100G
APOSOR963214-25GEA 270 EUR
APOSOR963214-25G APOSOR963214-5G APOSOR963214-100G
Ac-Nva-OH 98%
Ac-Nva-OH
Formula: C₇H₁₃NO₃
Storage Temperature: Ambient
CAS Number: 15891-50-6