Naphthalene-2,3-dicarbonitrile 98%
Supplier: Apollo Scientific
Synonyms: 2,3-Naphthalenedicarbonitrile, 2,3-Dicyanonaphthalene
OR963527-1G OR963527-5G
APOSOR963527-1GEA 63.8 EUR
APOSOR963527-1G APOSOR963527-5G
Naphthalene-2,3-dicarbonitrile 98%
Naphthalene-2,3-dicarbonitrile
Formula: C₁₂H₆N₂
MW: 178.1894 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059063
CAS Number: 22856-30-0