3-(4-Methoxyphenyl)propan-1-amine 97%
Supplier: Apollo Scientific
OR964300-1G OR964300-5G
APOSOR964300-1GEA 116 EUR
APOSOR964300-1G APOSOR964300-5G
3-(4-Methoxyphenyl)propan-1-amine 97%
3-(4-Methoxyphenyl)propan-1-amine
Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00870510
CAS Number: 36397-23-6