N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine hydrochloride
Supplier: Apollo Scientific
Synonyms: 1-Methyl-2-[(methylamino)methyl]-1H-benzimidazole
OR9671-1G OR9671-250MG
APOSOR9671-1GEA 352 EUR
APOSOR9671-1G APOSOR9671-250MG
N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine hydrochloride
N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine hydrochloride
Formula: C₁₀H₁₄ClN₃
MW: 211.7 g/mol
Melting Pt: 81.5…84.5 °C
Storage Temperature: Ambient
MDL Number: MFCD12027123
CAS Number: 137898-62-5