2,5-Dibromobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,5-Dibromobenzene-1-sulphonyl chloride
OR9709-5G OR9709-1G
APOSOR9709-5GEA 0 EUR
APOSOR9709-5G APOSOR9709-1G
2,5-Dibromobenzenesulphonyl chloride 97%
2,5-Dibromobenzenesulphonyl chloride
Formula: C₆H₃Br₂ClO₂S
MW: 334.42 g/mol
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00060685
CAS Number: 23886-64-8
UN: 3261
ADR: 8 II