Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 2-chloro-4-methylthiazole-5-carboxylate
OR9735-1G OR9735-250MG
APOSOR9735-1GEA 21.9 EUR
APOSOR9735-1G APOSOR9735-250MG
Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate 97%
Ethyl-2-chloro-4-methyl-1,3-thiazole-5-carboxylate
Formula: C₇H₈ClNO₂S
MW: 205.66 g/mol
Melting Pt: 46…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02956798
CAS Number: 7238-62-2