2-(2-Pyridyl)malondialdehyde
Supplier: Apollo Scientific
Warning
OR9773-1G OR9773-250MG
APOSOR9773-1GEA 72.5 EUR
APOSOR9773-1G APOSOR9773-250MG
2-(2-Pyridyl)malondialdehyde
2-(2-Pyridyl)malondialdehyde
Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 202…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00216529
CAS Number: 212755-83-4