4-Bromobenzyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene
OR9776-25G OR9776-5G
APOSOR9776-25GEA 50.9 EUR
APOSOR9776-25G APOSOR9776-5G
4-Bromobenzyl chloride
4-Bromobenzyl chloride
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 106…107 °C/10mmHg
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040867
CAS Number: 589-17-3
EINECS: 209-638-6
UN: 3261
ADR: 8 II
Merck Index: 13,01394