3-[(3R,4R)-3,4-Dimethylpiperidin-4-yl]phenol
Supplier: Apollo Scientific
Synonyms: (3R,4R)-3,4-Dimethyl-4-(3-hydroxyphenyl)piperidine
OR9962-250MG OR9962-50MG
APOSOR9962-250MGEA 150 EUR
APOSOR9962-250MG APOSOR9962-50MG
3-[(3R,4R)-3,4-Dimethylpiperidin-4-yl]phenol
3-[(3R,4R)-3,4-Dimethylpiperidin-4-yl]phenol
Formula: C₁₃H₁₉NO
MW: 205.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659960
CAS Number: 119193-19-0