2-Toluenethiol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Thiocresol, 1-Mercapto-2-methylbenzene, o-Toluenethiol, 2-Methylbenzene-1-thiol, 2-Methylbenzenethiol, o-Thiocresol, 2-Methylthiophenol
OR9990-10G OR9990-25G OR9990-100G
APOSOR9990-10GEA 0 EUR
APOSOR9990-10G APOSOR9990-25G APOSOR9990-100G
2-Toluenethiol 98%
2-Toluenethiol
Formula: C₇H₈S
Boiling Pt: 195 °C
Melting Pt: 44540 °C
Density: 1.054 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004838
CAS Number: 137-06-4
EINECS: 205-276-8
UN: 2810
ADR: 6.1,III