2-Chloro-3,6-difluorophenol 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Choro-3,6-difluorophenol
PC0012-25G PC0012-5G PC0012-250MG PC0012-1G
APOSPC0012-25GEA 585 EUR
APOSPC0012-25G APOSPC0012-5G APOSPC0012-250MG APOSPC0012-1G
2-Chloro-3,6-difluorophenol 97%
2-Chloro-3,6-difluorophenol
Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD01631450
CAS Number: 261762-50-9