3-Chloro-2-fluorobenzaldehyde 90%
Supplier: Apollo Scientific
Warning
PC0028-100G PC0028-25G PC0028-5G
APOSPC0028-100GEA 111 EUR
APOSPC0028-100G APOSPC0028-25G APOSPC0028-5G
3-Chloro-2-fluorobenzaldehyde 90%
3-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
Boiling Pt: 214 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD01631571
CAS Number: 85070-48-0